siRNA Details
| Gene Symbol: Gm36536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158658.1 | - | 4 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm36536| Gene Symbol: Gm36536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158658.1 | - | 4 | 882 | ||