siRNA Details
| Gene Symbol: BC016548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096650.1 | - | 3 | 461 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI117904.1 | - | 2 | 391 | Chr. 2 - 103377796 - 103388286 [-] on Build GRCm38 | |

BC016548
Genomic Map