siRNA Details
| Gene Symbol: Toporsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006061.1 | - | 1 | 2090 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB679288.1 | - | 1 | 247 | Chr. 4 - 52612102 - 52612427 [+] on Build GRCm38 | |

Toporsl
Genomic Map