siRNA Details
| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045079.1 | 1 | 384 | |||
| NR_045080.1 | 1 | 384 | ||||
| GenBank mRNA | AK019455.1 | - | 1 | 223 | ||
| AK077272.1 | - | 1 | 386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3300005D01Rik| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045079.1 | 1 | 384 | |||
| NR_045080.1 | 1 | 384 | ||||
| GenBank mRNA | AK019455.1 | - | 1 | 223 | ||
| AK077272.1 | - | 1 | 386 | |||