siRNA Details
| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045079.1 | 1 | 240 | |||
| NR_045080.1 | 1 | 240 | ||||
| GenBank mRNA | AK019455.1 | - | 1 | 79 | ||
| AK077272.1 | - | 1 | 242 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713361.1 | - | 1 | 76 | Chr. 17 - 5799654 - 5802897 [+] on Build GRCm38 | |

3300005D01Rik
Genomic Map