siRNA Details
| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045079.1 | 2 | 607 | |||
| NR_045081.1 | 2 | 207 | ||||
| GenBank mRNA | AA008373.1 | - | 2 | 210 | ||
| AK077272.1 | - | 2 | 609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3300005D01Rik| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045079.1 | 2 | 607 | |||
| NR_045081.1 | 2 | 207 | ||||
| GenBank mRNA | AA008373.1 | - | 2 | 210 | ||
| AK077272.1 | - | 2 | 609 | |||