siRNA Details
| Gene Symbol: 1700017G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040445.1 | 3 | 697 | |||
| GenBank mRNA | AK006064.1 | - | 3 | 698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700017G19Rik| Gene Symbol: 1700017G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040445.1 | 3 | 697 | |||
| GenBank mRNA | AK006064.1 | - | 3 | 698 | ||