siRNA Details
| Gene Symbol: 1700017I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006067.1 | - | 2 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706181.1 | - | 2 | 252 | Chr. 9 - 89802870 - 89806055 [+] on Build GRCm38 | |

1700017I07Rik
Genomic Map