siRNA Details
| Gene Symbol: E230013L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054035.1 | - | 1 | 733 | ||
| BC030398.1 | - | 1 | 710 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230013L22Rik| Gene Symbol: E230013L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054035.1 | - | 1 | 733 | ||
| BC030398.1 | - | 1 | 710 | |||