siRNA Details
| Gene Symbol: 4930591A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019836.1 | - | 3 | 346 | ||
| AK019837.1 | - | 3 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930591A17Rik| Gene Symbol: 4930591A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019836.1 | - | 3 | 346 | ||
| AK019837.1 | - | 3 | 356 | |||