siRNA Details
| Gene Symbol: Fendrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087264.1 | - | 6 | 2344 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB534926.2 | - | 2 | 624 | Chr. 8 - 121054882 - 121059974 [-] on Build GRCm38 | |

Fendrr
Genomic Map