siRNA Details
| Gene Symbol: 0610012D04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002586.1 | - | 2 | 148 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY754649.1 | - | 2 | 148 | Chr. 17 - 87362242 - 87364462 [-] on Build GRCm38 | |

0610012D04Rik
Genomic Map