siRNA Details
| Gene Symbol: Gm34481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033826.1 | - | 191 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB625590.1 | - | 2 | 191 | Chr. 10 - 43659318 - 43660753 [+] on Build GRCm38 | |

Gm34481
Genomic Map