siRNA Details
| Gene Symbol: Gm9873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079848.1 | - | 1 | 792 | ||
| BE955693.1 | - | 1 | 387 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB209350.2 | - | 1 | 332 | Chr. 2 - 169020731 - 169021366 [+] on Build GRCm38 | |

Gm9873
Genomic Map