siRNA Details
| Gene Symbol: 1700025M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040687.1 | 5 | 713 | |||
| NR_040688.1 | 4 | 621 | ||||
| NR_040689.1 | 3 | 541 | ||||
| GenBank mRNA | AK006359.1 | - | 3 | 544 | ||
| BB014226.1 | - | 1 | 209 | |||
| BC048655.1 | - | 5 | 710 | |||

1700025M24Rik
Genomic Map