siRNA Details
| Gene Symbol: Gm15326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134426.1 | - | 3 | 606 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM461161.1 | - | 3 | 576 | Chr. 13 - 111867974 - 111871585 [+] on Build GRCm38 | |

Gm15326
Genomic Map