siRNA Details
| Gene Symbol: 2310007H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009208.1 | - | 1 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709017.1 | - | 1 | 596 | Chr. 2 - 164522215 - 164523044 [+] on Build GRCm38 | |

2310007H11Rik
Genomic Map