siRNA Details
Gene Symbol: 4921525O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045061.1 | 7 | 2017 | |||
GenBank mRNA | AK014969.1 | - | 7 | 2018 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921525O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045061.1 | 7 | 2017 | |||
GenBank mRNA | AK014969.1 | - | 7 | 2018 |