siRNA Details
| Gene Symbol: 4921525O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045061.1 | 7 | 1834 | |||
| GenBank mRNA | AK014969.1 | - | 7 | 1835 | ||
| DQ563489.1 | - | 1 | 12 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921525O09Rik| Gene Symbol: 4921525O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045061.1 | 7 | 1834 | |||
| GenBank mRNA | AK014969.1 | - | 7 | 1835 | ||
| DQ563489.1 | - | 1 | 12 | |||