siRNA Details
| Gene Symbol: 4930473O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045356.1 | 1 | 336 | |||
| GenBank mRNA | AK015568.1 | - | 1 | 337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930473O22Rik| Gene Symbol: 4930473O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045356.1 | 1 | 336 | |||
| GenBank mRNA | AK015568.1 | - | 1 | 337 | ||