siRNA Details
Gene Symbol: 4930473O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045356.1 | 3 | 1450 | |||
GenBank mRNA | AK015568.1 | - | 3 | 1451 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930473O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045356.1 | 3 | 1450 | |||
GenBank mRNA | AK015568.1 | - | 3 | 1451 |