siRNA Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008866.1 | - | 1 | 612 | ||
| AK141895.1 | - | 1 | 1829 | |||
| AK151977.1 | - | 1 | 1358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc44a1| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008866.1 | - | 1 | 612 | ||
| AK141895.1 | - | 1 | 1829 | |||
| AK151977.1 | - | 1 | 1358 | |||