siRNA Details
| Gene Symbol: Gm26761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033648.1 | - | 1 | 34 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB625243.1 | - | 1 | 34 | Chr. 5 - 64696987 - 64698520 [-] on Build GRCm38 | |

Gm26761
Genomic Map