siRNA Details
| Gene Symbol: Zfp493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138937.1 | - | 3517 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB214508.2 | - | 1 | 129 | Chr. 13 - 67789346 - 67789916 [+] on Build GRCm38 | |

Zfp493
Genomic Map