siRNA Details
| Gene Symbol: 4930503O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040477.1 | 1 | 204 | |||
| GenBank mRNA | AK015691.1 | - | 1 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715232.1 | - | 1 | 205 | Chr. 1 - 194212187 - 194222759 [-] on Build GRCm38 | |

4930503O07Rik
Genomic Map