siRNA Details
| Gene Symbol: Gm6639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040748.1 | 8 | 820 | |||
| GenBank mRNA | AK029958.1 | - | 8 | 820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6639| Gene Symbol: Gm6639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040748.1 | 8 | 820 | |||
| GenBank mRNA | AK029958.1 | - | 8 | 820 | ||