siRNA Details
| Gene Symbol: Gm31219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136039.1 | - | 1, 2 | 185 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719968.1 | - | 1, 2 | 182 | Chr. 13 - 76255949 - 76270964 [+] on Build GRCm38 | |

Gm31219
Genomic Map