siRNA Details
| Gene Symbol: 1700109G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046197.1 | 1 | 35 | |||
| GenBank mRNA | AK018946.1 | - | 1 | 36 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707234.1 | - | 1 | 36 | Chr. 11 - 84716975 - 84720432 [-] on Build GRCm38 | |

1700109G15Rik
Genomic Map