siRNA Details
| Gene Symbol: 4933425D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007161.1 | - | 1 | 1248 | ||
| AK077189.1 | - | 1 | 1284 | |||
| AV206338.1 | - | 1 | 231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933425D22Rik| Gene Symbol: 4933425D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007161.1 | - | 1 | 1248 | ||
| AK077189.1 | - | 1 | 1284 | |||
| AV206338.1 | - | 1 | 231 | |||