siRNA Details
| Gene Symbol: Gm16070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038933.1 | - | 4, 5 | 617 | ||
| AK157774.1 | - | 3, 4 | 912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16070| Gene Symbol: Gm16070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038933.1 | - | 4, 5 | 617 | ||
| AK157774.1 | - | 3, 4 | 912 | |||