siRNA Details
| Gene Symbol: 4930578L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028578.1 | - | 1 | 2894 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV235277.2 | - | 1 | 279 | Chr. 18 - 73706551 - 73707209 [+] on Build GRCm38 | |

4930578L24Rik
Genomic Map