siRNA Details
Gene Symbol: 1700028P15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040509.1 | 6 | 588 | |||
GenBank mRNA | AK006474.1 | - | 6 | 589 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706513.1 | - | 6 | 589 | Chr. 2 - 171957031 - 171962799 [-] on Build GRCm38 |
