siRNA Details
| Gene Symbol: 1700028P15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040509.1 | 2 | 83 | |||
| GenBank mRNA | AK006474.1 | - | 2 | 84 | ||
| AK133337.1 | - | 1 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700028P15Rik| Gene Symbol: 1700028P15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040509.1 | 2 | 83 | |||
| GenBank mRNA | AK006474.1 | - | 2 | 84 | ||
| AK133337.1 | - | 1 | 229 | |||