siRNA Details
| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053027.1 | - | 4 | 1863 | ||
| AK144771.1 | - | 4 | 1655 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15834| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053027.1 | - | 4 | 1863 | ||
| AK144771.1 | - | 4 | 1655 | |||