siRNA Details
Gene Symbol: 4933439K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040558.1 | 3 | 739 | |||
GenBank mRNA | AK017125.1 | - | 3 | 741 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933439K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040558.1 | 3 | 739 | |||
GenBank mRNA | AK017125.1 | - | 3 | 741 |