siRNA Details
| Gene Symbol: 4933439K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040558.1 | 2, 3 | 397 | |||
| GenBank mRNA | AK017125.1 | - | 2, 3 | 399 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716803.1 | - | 397 | Chr. - - [-] on Build | ||

4933439K11Rik
Genomic Map