siRNA Details
| Gene Symbol: 4933439K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040558.1 | 3 | 776 | |||
| GenBank mRNA | AK017125.1 | - | 3 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933439K11Rik| Gene Symbol: 4933439K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040558.1 | 3 | 776 | |||
| GenBank mRNA | AK017125.1 | - | 3 | 778 | ||