siRNA Details
| Gene Symbol: 2810427C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013171.1 | - | 1 | 33 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712193.1 | - | 1 | 39 | Chr. 2 - 92306175 - 92307120 [+] on Build GRCm38 | |

2810427C15Rik
Genomic Map