siRNA Details
| Gene Symbol: 2810427C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013171.1 | - | 1 | 723 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712193.1 | - | 1 | 729 | Chr. 2 - 92306175 - 92307120 [+] on Build GRCm38 | |
 Back To Top
 Back To Top
 2810427C15Rik
2810427C15Rik Genomic Map
Genomic Map Back To Top
 Back To Top