siRNA Details
| Gene Symbol: 4933402C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045504.1 | 4 | 729 | |||
| GenBank mRNA | AK016610.1 | - | 4 | 730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933402C06Rik| Gene Symbol: 4933402C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045504.1 | 4 | 729 | |||
| GenBank mRNA | AK016610.1 | - | 4 | 730 | ||