siRNA Details
| Gene Symbol: 1700095A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007073.1 | - | 638 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707142.1 | - | 2 | 639 | Chr. 18 - 83106528 - 83107830 [-] on Build GRCm38 | |

1700095A13Rik
Genomic Map