siRNA Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007077.1 | - | 1 | 41 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707148.1 | - | 1 | 41 | Chr. 16 - 33966239 - 33967312 [+] on Build GRCm38 | |

Umps
Genomic Map