siRNA Details
| Gene Symbol: 1700024F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045363.1 | 2 | 170 | |||
| GenBank mRNA | AK006297.1 | - | 2 | 179 | ||
| Gene Symbol: 2810429I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013191.1 | - | 636 | |||

1700024F13Rik
Genomic Map