siRNA Details
Gene Symbol: 1700024F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045363.1 | 3 | 433 | |||
GenBank mRNA | AK006297.1 | - | 3 | 442 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700024F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045363.1 | 3 | 433 | |||
GenBank mRNA | AK006297.1 | - | 3 | 442 |