siRNA Details
| Gene Symbol: Gm6145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045385.1 | - | 2437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6145| Gene Symbol: Gm6145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045385.1 | - | 2437 | |||