siRNA Details
| Gene Symbol: 9230111E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020334.1 | - | 1 | 21 | ||
| BY721144.1 | - | 1 | 21 | |||
| Gene Symbol: Gm14321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CK619576.1 | - | 4 | 578 | ||

9230111E07Rik
Genomic Map