siRNA Details
| Gene Symbol: 9430076D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020488.1 | - | 1 | 242 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721851.1 | - | 1 | 242 | Chr. 16 - 38527732 - 38528416 [+] on Build GRCm38 | |

9430076D03Rik
Genomic Map