siRNA Details
Gene Symbol: 9430076D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020488.1 | - | 1 | 150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721851.1 | - | 1 | 150 | Chr. 16 - 38527732 - 38528416 [+] on Build GRCm38 |
