siRNA Details
| Gene Symbol: Redrum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040338.1 | 3 | 1916 | |||
| GenBank mRNA | AK020489.1 | - | 1 | 699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Redrum| Gene Symbol: Redrum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040338.1 | 3 | 1916 | |||
| GenBank mRNA | AK020489.1 | - | 1 | 699 | ||