siRNA Details
| Gene Symbol: 6720468P15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040306.1 | 3 | 670 | |||
| NR_040307.1 | 2 | 308 | ||||
| GenBank mRNA | AK020151.1 | - | 3 | 648 | ||
| AK040090.1 | - | 3 | 611 | |||
| AK078497.1 | - | 3 | 671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6720468P15Rik| Gene Symbol: 6720468P15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040306.1 | 3 | 670 | |||
| NR_040307.1 | 2 | 308 | ||||
| GenBank mRNA | AK020151.1 | - | 3 | 648 | ||
| AK040090.1 | - | 3 | 611 | |||
| AK078497.1 | - | 3 | 671 | |||